Assay Details
| Gene Symbol: Lrrc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142728.1 | NP_001136200.1 | 3 - 4 | 605 | 173 | |
| NM_001142729.1 | NP_001136201.1 | 3 - 4 | 524 | 173 | ||
| NM_030070.3 | NP_084346.2 | 3 - 4 | 605 | 173 | ||
| GenBank mRNA | AK015292.1 | - | 3 - 4 | 523 | 173 | |
| AK019545.1 | - | 3 - 4 | 538 | 173 | ||
| AK029802.1 | - | 3 - 4 | 606 | 173 | ||
| AK031279.1 | - | 3 - 4 | 524 | 173 | ||
| AK039037.1 | - | 3 - 4 | 415 | 173 | ||
| AK082549.1 | - | 3 - 4 | 532 | 173 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc9
Genomic Map