Assay Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172862.3 | NP_766450.2 | 2 - 3 | 5500 | 80 | |
| GenBank mRNA | AB236662.1 | - | 2 - 3 | 5500 | 80 | |
| AJ833643.1 | - | 2 - 3 | 5229 | 80 | ||
| CN534900.1 | - | 1 - 2 | 63 | 80 | ||
| CN536459.1 | - | 2 - 3 | 165 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frem2
Genomic Map