| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300015077.mm.1 |
NM_172862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015077.mm.1 |
NM_172862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015077.mm.1 |
TC0300001983.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300015078.mm.1 |
NM_172862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015078.mm.1 |
NM_172862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015078.mm.1 |
TC0300001983.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300015076.mm.1 |
NM_172862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015076.mm.1 |
NM_172862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015076.mm.1 |
TC0300001983.mm.1 |
MTA TC |