Assay Details
| Gene Symbol: Agrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021604.3 | NP_067617.3 | 22 - 23 | 3795 | 64 | |
| GenBank mRNA | AK147577.1 | - | 20 - 21 | 4228 | 64 | |
| BC059259.1 | - | 9 - 10 | 1209 | 64 | ||
| BC150703.1 | - | 20 - 21 | 3249 | 9087614 | 64 | |
More Information
Set Membership: |
|

Agrn
Genomic Map