Assay Details
| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF513723.1 | - | 13 - 14 | 2598 | 70 | |
| AF513724.1 | - | 12 - 13 | 2409 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Disc1| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF513723.1 | - | 13 - 14 | 2598 | 70 | |
| AF513724.1 | - | 12 - 13 | 2409 | 70 | ||
Set Membership: |
|