Assay Details
| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174854.2 | NP_777279.2 | 9 - 10 | 2008 | 107 | |
| GenBank mRNA | AY177673.1 | - | 9 - 10 | 1973 | 107 | |
| AY320287.1 | - | 10 - 11 | 2090 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Disc1
Genomic Map