Assay Details
| Gene Symbol: Twf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008971.4 | NP_032997.3 | 6 - 7 | 655 | 117 | |
| GenBank mRNA | AK077410.1 | - | 6 - 7 | 714 | 117 | |
| AK147990.1 | - | 6 - 7 | 649 | 117 | ||
| AK150852.1 | - | 6 - 7 | 678 | 117 | ||
| AK168142.1 | - | 6 - 7 | 673 | 117 | ||
| AK168161.1 | - | 6 - 7 | 673 | 117 | ||
| AY267188.1 | - | 6 - 7 | 607 | 117 | ||
| BC015081.1 | - | 6 - 7 | 672 | 3979221 | 117 | |
| BC094034.1 | - | 6 - 7 | 655 | 4934593 | 117 | |
| U82324.1 | - | 6 - 7 | 663 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Twf1
Genomic Map