Assay Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026032.1 | NP_080308.1 | 3 - 4 | 314 | 67 | |
| GenBank mRNA | AK006006.1 | - | 1 - 2 | 49 | 67 | |
| AK012814.1 | - | 1 - 2 | 49 | 67 | ||
| AK019398.1 | - | 1 - 2 | 44 | 67 | ||
| AK041290.1 | - | 3 - 4 | 314 | 67 | ||
| BC021460.1 | - | 3 - 4 | 597 | 5054997 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
deadenylation-dependent decapping of nuclear-transcribed mRNA
mRNA processing
RNA splicing
stem cell population maintenance
negative regulation of neuron differentiation
histone mRNA catabolic process
RNA cap binding
RNA binding
mRNA binding
pre-mRNA binding
poly(A) RNA binding
deadenylation-dependent decapping of nuclear-transcribed mRNA
mRNA processing
RNA splicing
stem cell population maintenance
negative regulation of neuron differentiation
histone mRNA catabolic process
RNA cap binding
RNA binding
mRNA binding
pre-mRNA binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25729011 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25729014 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23678 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109533 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169049 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109533 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06454961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169049 | - | - |
| Whole Mouse Genome G4122A | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | AK041290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | BC021460.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | NM_026032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | AK006006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206655 | AK012814.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109533 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07215940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07215938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25729014 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25729011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25729093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07121743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06743176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07177766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07350917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07270991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07198789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07215938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07177764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07267034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25729079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07177767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07069639 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10571049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571053 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571052 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571051 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423873_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441452_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10571048 | - | - |
| Mouse Genome 430 2.0 Array | 1423873_at | - | - |
| Mouse Genome 430 2.0 Array | 1423873_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441452_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441452_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67207_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67207_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800002494.mm.1 | NM_026032.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002494.mm.1 | NM_026032.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002494.mm.1 | TC0800000330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800002495.mm.1 | NM_026032.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002495.mm.1 | NM_026032.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002495.mm.1 | TC0800000330.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423873_at | - | - |
| Mouse Expression Set 430 | 1423873_PM_at | - | - |
| Mouse Expression Set 430 | 1441452_at | - | - |
| Mouse Expression Set 430 | 1441452_PM_at | - | - |
| Murine Genome U74v2 Set | 96946_PM_at | - | - |
| Murine Genome U74v2 Set | 96946_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423873_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423873_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423873_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423873_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441452_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441452_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 924957 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106370390 | - | - |
| MouseWG-6 v2 Expression BeadChip | 100450170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106370286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103710008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106370390 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226536 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2603984 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244979 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2568554 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2556157 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103710008 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106370286 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106370390 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106370390 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103710008 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106370286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2603984 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2919270 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244979 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2568554 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2556157 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2919270 | - | - |

Lsm1
Genomic Map