Assay Details
| Gene Symbol: Igfals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020357.1 | - | 1 - 2 | 76 | 73 | |
| BC106111.1 | - | 1 - 2 | 59 | 73 | ||
| BC129875.1 | - | 1 - 2 | 44 | 40110389 | 73 | |
| BC129876.1 | - | 1 - 2 | 44 | 40110391 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC106111.1, BC129875.1, BC129876.1 transcripts.
Set Membership: |
|

Igfals
Genomic Map