Assay Details
| Gene Symbol: Krt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010664.2 | NP_034794.2 | 1 - 2 | 458 | 69 | |
| GenBank mRNA | AK145413.1 | - | 1 - 2 | 448 | 69 | |
| AK167072.1 | - | 1 - 2 | 447 | 69 | ||
| AK167265.1 | - | 1 - 2 | 442 | 69 | ||
| AK167432.1 | - | 1 - 2 | 445 | 69 | ||
| AK167469.1 | - | 1 - 2 | 448 | 69 | ||
| AK167676.1 | - | 1 - 2 | 447 | 69 | ||
| BC020474.1 | - | 1 - 2 | 448 | 69 | ||
| BC089022.1 | - | 1 - 2 | 420 | 30701486 | 69 | |
| CV561123.1 | - | 1 - 2 | 460 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt18
Genomic Map