Assay Details
| Gene Symbol: Cmpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025647.3 | NP_079923.3 | 4 - 5 | 636 | 113 | |
| GenBank mRNA | AK002526.1 | - | 4 - 5 | 555 | 113 | |
| AK004827.1 | - | 4 - 5 | 525 | 113 | ||
| AK077534.1 | - | 4 - 5 | 651 | 113 | ||
| AK146436.1 | - | 4 - 5 | 558 | 113 | ||
| AK150162.1 | - | 4 - 5 | 582 | 113 | ||
| AK161967.1 | - | 219 | 113 | |||
| AK162173.1 | - | 4 - 5 | 580 | 113 | ||
| BC017684.1 | - | 4 - 5 | 564 | 4241782 | 113 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cmpk1
Genomic Map