Assay Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191004.1 | NP_001177933.1 | 2 - 3 | 201 | 86 | |
| GenBank mRNA | AK019126.1 | - | 2 - 3 | 161 | 86 | |
| AK078392.1 | - | 2 - 3 | 151 | 86 | ||
| AK141884.1 | - | 2 - 3 | 148 | 86 | ||
| AK161686.1 | - | 2 - 3 | 155 | 86 | ||
| BC099484.1 | - | 1 - 2 | 107 | 30936207 | 86 | |
| DV648841.1 | - | 2 - 3 | 201 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019126.1, AK078392.1, AK141884.1, AK161686.1, BC099484.1, NM_001191004.1 transcripts.
Set Membership: |
|

Lsm6
Genomic Map