| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10583786 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10583787 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1452201_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10583785 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10591658 |
- |
- |
| Mouse Genome 430 2.0 Array |
1452201_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1431442_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1431442_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1452201_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66962_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66962_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900001591.mm.1 |
NM_025870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001591.mm.1 |
NM_025870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001591.mm.1 |
TC0900000209.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001590.mm.1 |
NM_025870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001590.mm.1 |
NM_025870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001590.mm.1 |
TC0900000209.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001589.mm.1 |
NM_025870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001589.mm.1 |
NM_025870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001589.mm.1 |
TC0900000209.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1452201_PM_at |
- |
- |
| Mouse Expression Set 430 |
1452201_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1452201_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1452201_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1452201_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1452201_PM_at |
- |
- |