Assay Details
| Gene Symbol: Tdgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011562.2 | NP_035692.2 | 1 - 2 | 339 | 92 | |
| GenBank mRNA | AK133730.1 | - | 1 - 2 | 277 | 92 | |
| AK163969.1 | - | 1 - 2 | 276 | 92 | ||
| BC052646.1 | - | 1 - 2 | 281 | 30029798 | 92 | |
| CJ074826.1 | - | 1 - 2 | 339 | 92 | ||
| M87321.1 | - | 1 - 2 | 248 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133730.1,BC052646.1,M87321.1,NM_011562.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tdgf1
Genomic Map