Assay Details
| Gene Symbol: Tdgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011562.2 | NP_035692.2 | 2 - 3 | 390 | 74 | |
| GenBank mRNA | AK133730.1 | - | 2 - 3 | 328 | 74 | |
| AK163969.1 | - | 2 - 3 | 327 | 74 | ||
| BC052646.1 | - | 2 - 3 | 332 | 30029798 | 74 | |
| M87321.1 | - | 2 - 3 | 299 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tdgf1
Genomic Map