Assay Details
| Gene Symbol: Tdgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011562.2 | NP_035692.2 | 4 - 5 | 607 | 62 | |
| GenBank mRNA | AK133730.1 | - | 4 - 5 | 545 | 62 | |
| BC052646.1 | - | 4 - 5 | 549 | 30029798 | 62 | |
| M87321.1 | - | 4 - 5 | 516 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
vasculogenesis
in utero embryonic development
morphogenesis of a branching structure
positive regulation of cell-matrix adhesion
cell migration involved in sprouting angiogenesis
epidermal growth factor receptor signaling pathway
gastrulation
heart development
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
anterior/posterior axis specification
regulation of signal transduction
positive regulation of endothelial cell migration
peptidyl-serine phosphorylation
cell differentiation
regulation of cell migration
positive regulation of cell migration
BMP signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
cellular response to hepatocyte growth factor stimulus
nodal signaling pathway
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
positive regulation of peptidyl-tyrosine phosphorylation
cardiac muscle cell differentiation
canonical Wnt signaling pathway
cellular response to interferon-gamma
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
receptor binding
protein binding
growth factor activity
coreceptor activity
Wnt-protein binding
nodal binding
type I activin receptor binding
activation of MAPK activity
vasculogenesis
in utero embryonic development
morphogenesis of a branching structure
positive regulation of cell-matrix adhesion
cell migration involved in sprouting angiogenesis
epidermal growth factor receptor signaling pathway
gastrulation
heart development
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
anterior/posterior axis specification
regulation of signal transduction
positive regulation of endothelial cell migration
peptidyl-serine phosphorylation
cell differentiation
regulation of cell migration
positive regulation of cell migration
BMP signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
cellular response to hepatocyte growth factor stimulus
nodal signaling pathway
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
positive regulation of peptidyl-tyrosine phosphorylation
cardiac muscle cell differentiation
canonical Wnt signaling pathway
cellular response to interferon-gamma
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
receptor binding
protein binding
growth factor activity
coreceptor activity
Wnt-protein binding
nodal binding
type I activin receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058540 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968028 | BC052646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968028 | NM_011562.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968028 | AK133730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528306 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145372 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183147 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058540 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968028 | BC052646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968028 | NM_011562.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968028 | AK133730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23716 | M87321.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826831 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145372 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02029916 | - | - |
| Whole Mouse Genome G4122A | A_52_P23716 | M87321.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124158 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145372 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07947581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07826099 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07947579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07947580 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07826101 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08131152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07506984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07544884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07820708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07945960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08141795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07723721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07640435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07947575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08064658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07947581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826758 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10597271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597270 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450989_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597268 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433197 | - | - |
| Mouse Genome 430 2.0 Array | 1450989_at | - | - |
| Mouse Genome 430 2.0 Array | 1450989_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21667_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21667_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900026435.mm.1 | NM_011562.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900026435.mm.1 | NM_011562.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900026435.mm.1 | TC0900003121.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900026436.mm.1 | NM_011562.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900026436.mm.1 | NM_011562.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900026436.mm.1 | TC0900003121.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450989_PM_at | - | - |
| Mouse Expression Set 430 | 1450989_at | - | - |
| Murine Genome U74v2 Set | 93002_r_at | - | - |
| Murine Genome U74v2 Set | 93002_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450989_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450989_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450989_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450989_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 482685 | - | - |
| ABI Mouse Genome Survey Microarray | 897869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1690338 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6760070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6760070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2595311 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697684 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697684 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2595311 | - | - |
| mouse-6 v1.1 (Illumina) | scl021667.1_237-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0021667.1_330-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690338 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0021667.1_330-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl021667.1_237-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690338 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2595311 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2917458 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2595311 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2917458 | - | - |

Tdgf1
Genomic Map