Assay Details
| Gene Symbol: Slc39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013901.2 | NP_038929.2 | 3 - 4 | 414 | 79 | |
| GenBank mRNA | AK146175.1 | - | 3 - 4 | 406 | 79 | |
| AK159290.1 | - | 3 - 4 | 271 | 79 | ||
| BC005474.1 | - | 3 - 4 | 376 | 3591856 | 79 | |
| BC132461.1 | - | 2 - 3 | 351 | 40130738 | 79 | |
| BC138459.1 | - | 2 - 3 | 351 | 8861479 | 79 | |
| BC147738.1 | - | 2 - 3 | 352 | 9056687 | 79 | |
| BC147744.1 | - | 2 - 3 | 352 | 9056693 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc39a1
Genomic Map