Assay Details
| Gene Symbol: Olfr1354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199840.1 | NP_001186769.1 | 1 | 629 | 143 | |
| GenBank mRNA | CB173515.1 | - | 1 | 231 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1354
Genomic Map