Assay Details
| Gene Symbol: Tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024214.2 | NP_077176.1 | 3 - 4 | 448 | 86 | |
| GenBank mRNA | AK002902.1 | - | 3 - 4 | 369 | 86 | |
| AK160168.1 | - | 3 - 4 | 351 | 86 | ||
| AK181461.1 | - | 3 - 4 | 167 | 86 | ||
| AK185977.1 | - | 167 | 86 | |||
| BC002087.1 | - | 3 - 4 | 357 | 3484983 | 86 | |
| BC099449.1 | - | 3 - 4 | 355 | 30848526 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26260529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26260532 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26260534 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136338 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94964 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127096 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137810 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041017 | AK160168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2136338 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26260513 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127096 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137810 | - | - |
| Whole Mouse Genome G4122A | A_52_P310930 | BC099449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310930 | AK141367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310930 | AK160168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310930 | AK181461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310930 | AK185977.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143457 | AK160168.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107768 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137810 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07379781 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07231080 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07379776 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045311 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045315 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045313 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07231075 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26260532 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26260534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26260529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26260536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07026818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07231074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06939661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07223522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07223509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07231075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06763367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07282145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07223527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06751111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07023862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26260519 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10582820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477371 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455357_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423080_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435534_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423081_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10582814 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477370 | - | - |
| Mouse Genome 430 2.0 Array | 1435534_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423080_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423081_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423080_at | - | - |
| Mouse Genome 430 2.0 Array | 1435187_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423081_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423079_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435187_at | - | - |
| Mouse Genome 430 2.0 Array | 1455357_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1435534_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423079_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455357_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800024616.mm.1 | NM_024214.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024616.mm.1 | NM_024214.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024616.mm.1 | TC0800003200.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423079_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423079_a_at | - | - |
| Mouse Expression Set 430 | 1455357_x_at | - | - |
| Mouse Expression Set 430 | 1435534_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423081_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423080_at | - | - |
| Mouse Expression Set 430 | 1423080_PM_at | - | - |
| Mouse Expression Set 430 | 1455357_PM_x_at | - | - |
| Mouse Expression Set 430 | 1423081_a_at | - | - |
| Mouse Expression Set 430 | 1435534_a_at | - | - |
| Murine Genome U74v2 Set | 101036_PM_at | - | - |
| Murine Genome U74v2 Set | 101036_at | - | - |
| Murine Genome U74v2 Set | 171205_at | - | - |
| Murine Genome U74v2 Set | 171205_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423081_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455357_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423079_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435534_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423080_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423081_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455357_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423080_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423079_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435534_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435534_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455357_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435534_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423081_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455357_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423081_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 893672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6510113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006510113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2477259 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2477259 | - | - |
| mouse-6 v1.1 (Illumina) | scl067952.2_217-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6510113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067952.2_217-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2477259 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2477259 | - | - |

Tomm20
Genomic Map