Assay Details
| Gene Symbol: Serpinb3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201363.2 | NP_958751.2 | 6 - 7 | 666 | 145 | |
| GenBank mRNA | AK003650.1 | - | 6 - 7 | 667 | 145 | |
| AY367775.1 | - | 5 - 6 | 596 | 145 | ||
| BC132091.1 | - | 5 - 6 | 618 | 40130368 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34792 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | NM_201363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P100997 | AK003650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P101004 | AK003650.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111347 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04217625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20512046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20512044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20512097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20512036 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10357093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357092 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357090 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100023514.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023514.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023514.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023512.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023512.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023512.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023513.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023513.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023513.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023517.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023517.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023517.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023516.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023516.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023516.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100031611.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031611.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031611.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023515.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023515.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023515.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023511.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023511.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023511.mm.1 | TC0100003885.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023510.mm.1 | NM_201363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023510.mm.1 | NM_201363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023510.mm.1 | TC0100003885.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422939_PM_at | - | - |
| Mouse Expression Set 430 | 1422939_at | - | - |
| Mouse Expression Set 430 | 1422940_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422940_x_at | - | - |
| Murine Genome U74v2 Set | 170906_r_at | - | - |
| Murine Genome U74v2 Set | 170908_f_at | - | - |
| Murine Genome U74v2 Set | 170908_PM_f_at | - | - |
| Murine Genome U74v2 Set | 170906_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171516_r_at | - | - |
| Murine Genome U74v2 Set | 171516_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422940_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422939_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422940_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422939_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 445923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2760600 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2760600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760600 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769409 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769409 | - | - |
| mouse-6 v1.1 (Illumina) | scl0381286.1_34-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760600 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0381286.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760600 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2824178 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2824178 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769409 | - | - |

Serpinb3c
Genomic Map