Assay Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011967.3 | NP_036097.1 | 1 - 2 | 381 | 167 | |
| GenBank mRNA | AK143468.1 | - | 1 - 2 | 383 | 167 | |
| AK160530.1 | - | 1 - 2 | 141 | 167 | ||
| AK160550.1 | - | 1 - 2 | 135 | 167 | ||
| AK160741.1 | - | 1 - 2 | 144 | 167 | ||
| AK161680.1 | - | 1 - 2 | 129 | 167 | ||
| BC007181.1 | - | 1 - 2 | 128 | 167 | ||
| BC010709.1 | - | 1 - 2 | 117 | 2648635 | 167 | |
| BC083342.1 | - | 1 - 2 | 114 | 5700963 | 167 | |
| BC108368.1 | - | 1 - 2 | 111 | 3596530 | 167 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143468.1,AK160530.1,AK160550.1,AK160741.1,AK161680.1,BC010709.1,BC083342.1,BC108368.1,NM_011967.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psma5
Genomic Map