| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1443944_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10507309 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10515335 |
- |
- |
| Mouse Genome 430 2.0 Array |
1443944_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1443944_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400026700.mm.1 |
TC0400003342.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026700.mm.1 |
NR_029450.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026700.mm.1 |
NR_029450.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026702.mm.1 |
TC0400003342.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026702.mm.1 |
NR_029450.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026702.mm.1 |
NR_029450.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026698.mm.1 |
TC0400003342.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026698.mm.1 |
NR_029450.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026698.mm.1 |
NR_029450.1 |
Refseq |
| Mouse Expression Set 430 |
1443944_at |
- |
- |
| Mouse Expression Set 430 |
1443944_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1443944_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1443944_at |
- |
- |