Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300024179.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024179.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024170.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024170.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024185.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024180.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024180.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024184.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024184.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024186.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024175.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024175.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024181.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024181.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024169.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024182.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024182.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024183.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024183.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024177.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024177.mm.1 |
TC0300003251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024178.mm.1 |
NM_009899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024178.mm.1 |
TC0300003251.mm.1 |
MTA TC |