Assay Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172097.1 | NP_001165568.1 | 6 - 7 | 1168 | 105 | |
| GenBank mRNA | AK019216.1 | - | 6 - 7 | 1102 | 105 | |
More Information
Set Membership: |
|

Cacul1
Genomic Map