Assay Details
| Gene Symbol: Serpinb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025429.2 | NP_079705.2 | 5 - 6 | 616 | 116 | |
| GenBank mRNA | AF426024.1 | - | 607 | 116 | ||
| AK003930.1 | - | 5 - 6 | 611 | 116 | ||
| AK076129.1 | - | 5 - 6 | 611 | 116 | ||
| AK160167.1 | - | 5 - 6 | 612 | 116 | ||
| BC011140.1 | - | 595 | 4207111 | 116 | ||
| BC104333.2 | - | 4 - 5 | 583 | 40045808 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb1a
Genomic Map