Assay Details
| Gene Symbol: Rpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048057.1 | NP_001041522.1 | 3 - 4 | 184 | 158 | |
| GenBank mRNA | AK010277.1 | - | 3 - 4 | 121 | 158 | |
| AK151517.1 | - | 3 - 4 | 121 | 158 | ||
| BC034339.1 | - | 3 - 4 | 123 | 3985781 | 158 | |
| CJ042718.1 | - | 3 - 4 | 158 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010277.1, AK151517.1, BC034339.1, NM_001048057.1 transcripts.
Set Membership: |
|

Rpl38
Genomic Map