Assay Details
| Gene Symbol: Rpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018860.4 | NP_061348.1 | 1 - 2 | 77 | 127 | |
| GenBank mRNA | BU937034.1 | - | 1 - 2 | 80 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_018860.4 transcripts.
Set Membership: |
|

Rpl41
Genomic Map