Assay Details
| Gene Symbol: Krt83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003668.2 | NP_001003668.2 | 4 - 5 | 778 | 177 | |
| GenBank mRNA | BC139321.1 | - | 4 - 5 | 778 | 8862343 | 177 |
| BC139322.1 | - | 4 - 5 | 778 | 8862344 | 177 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30491862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30491860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30491859 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966450 | BC139321.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966450 | NM_001003668.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966450 | BC139322.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105107 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170248 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966450 | BC139321.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966450 | NM_001003668.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966450 | BC139322.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30491863 | - | - |
| Whole Mouse Genome G4122A | A_52_P665240 | AK133718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P665240 | M92088.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P665240 | NM_001201323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P665240 | BC139321.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P665240 | NM_001003668.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117322 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139397 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12030520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12249235 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12467006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12030522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30491862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30491860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30491859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30491827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12404253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12125480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12435765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12179320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30491831 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432698 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432695 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500018552.mm.1 | NM_001003668.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018552.mm.1 | NM_001003668.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018552.mm.1 | TC1500002259.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018553.mm.1 | NM_001003668.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018553.mm.1 | NM_001003668.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018553.mm.1 | TC1500002259.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018554.mm.1 | NM_001003668.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018554.mm.1 | NM_001003668.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018554.mm.1 | TC1500002259.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2820127 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2820127 | - | - |

Krt83
Genomic Map