Assay Details
| Gene Symbol: Nme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008705.5 | NP_032731.1 | 1 - 2 | 427 | 149 | |
| GenBank mRNA | BC066995.1 | - | 1 - 2 | 427 | 5691909 | 149 |
| BY274368.1 | - | 1 - 2 | 178 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066995.1, NM_008705.5 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nme2
Genomic Map