Assay Details
| Gene Symbol: Rpl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009083.4 | NP_033109.1 | 1 - 2 | 200 | 130 | |
| GenBank mRNA | AK002988.1 | - | 1 - 2 | 198 | 130 | |
| BC069949.1 | - | 1 - 2 | 135 | 130 | ||
| CJ048578.1 | - | 1 - 2 | 201 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002988.1, NM_009083.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128575 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128574 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128582 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104941 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952475 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125437 | AK029389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125437 | CJ048578.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125437 | BC069949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125437 | AK002988.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125437 | NM_009083.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174481 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104941 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952475 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125437 | AK029389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125437 | CJ048578.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125437 | BC069949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125437 | AK002988.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125437 | NM_009083.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268914 | NR_028101.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268914 | BG312195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268914 | CO806526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268914 | BC069949.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268914 | AK002988.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128582 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174481 | - | - |
| Whole Mouse Genome G4122A | A_51_P268914 | NR_028101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268914 | BG312195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268914 | CO806526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268914 | BC069949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268914 | AK002988.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113373 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125437 | AK029389.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125437 | CJ048578.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125437 | BC069949.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125437 | AK002988.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125437 | NM_009083.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12201040 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12201037 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12201021 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12151615 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11860112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12183112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12201026 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128575 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128574 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30128581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11836782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11836781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12201030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12201021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12201022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12068452 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10397700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600028 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10428074 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600027 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10537349 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606255 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433562 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385570 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443193 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433902 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499198 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10397699 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606538 | - | - |
| Mouse Genome 430 2.0 Array | 1438076_at | - | - |
| Mouse Genome 430 2.0 Array | 1456266_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418273_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418273_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456266_at | - | - |
| Mouse Genome 430 2.0 Array | 1438076_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500010366.mm.1 | NM_001163485.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010366.mm.1 | NM_001163485.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010366.mm.1 | NM_009083.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010366.mm.1 | NM_009083.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010366.mm.1 | NR_028101.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010366.mm.1 | TC1500001345.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500010374.mm.1 | NM_009083.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010374.mm.1 | NM_009083.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010374.mm.1 | TC1500001345.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500010362.mm.1 | NM_001163485.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010362.mm.1 | NM_001163485.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010362.mm.1 | NM_009083.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010362.mm.1 | NM_009083.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010362.mm.1 | NR_028101.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010362.mm.1 | TC1500001345.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500010370.mm.1 | NM_001163485.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010370.mm.1 | NM_001163485.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010370.mm.1 | NM_009083.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010370.mm.1 | NM_009083.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010370.mm.1 | NR_028101.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010370.mm.1 | TC1500001345.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500010361.mm.1 | NM_001163485.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010361.mm.1 | NM_001163485.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010361.mm.1 | NM_009083.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010361.mm.1 | NM_009083.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010361.mm.1 | NR_028101.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010361.mm.1 | TC1500001345.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456266_PM_at | - | - |
| Mouse Expression Set 430 | 1418273_PM_a_at | - | - |
| Mouse Expression Set 430 | 1456266_at | - | - |
| Mouse Expression Set 430 | 1438076_at | - | - |
| Mouse Expression Set 430 | 1418273_a_at | - | - |
| Mouse Expression Set 430 | 1438076_PM_at | - | - |
| Murine Genome U74v2 Set | 98119_PM_at | - | - |
| Murine Genome U74v2 Set | 98119_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456266_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456266_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438076_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438076_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418273_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418273_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103610551 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4810369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103610551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810369 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2542887 | - | - |
| mouse-6 v1.1 (Illumina) | GI_42476346-S | - | - |
| mouse-6 v1.1 (Illumina) | scl019946.1_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103610551 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019946.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103610551 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2878430 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878424 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878430 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019946.1_13-S | - | - |

Rpl30
Genomic Map