Assay Details
| Gene Symbol: Nme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008704.2 | NP_032730.1 | 2 - 3 | 433 | 66 | |
| GenBank mRNA | AF033377.1 | - | 2 - 3 | 168 | 66 | |
| AK034899.1 | - | 172 | 66 | |||
| AK136353.1 | - | 2 - 3 | 248 | 66 | ||
| BC005629.1 | - | 2 - 3 | 172 | 3964056 | 66 | |
| M35970.1 | - | 2 - 3 | 170 | 66 | ||
| M65037.1 | - | 1 - 2 | 128 | 66 | ||
| U85511.1 | - | 2 - 3 | 220 | 66 | ||
More Information
Set Membership: |
|

Nme1
Genomic Map