Assay Details
Gene Symbol: Zfp114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029933.2 | NP_001025104.2 | 1 - 2 | 107 | 116 | |
GenBank mRNA | AF167317.1 | - | 1 - 2 | 92 | 116 | |
BB633884.1 | - | 1 - 2 | 107 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF167317.1, NM_001029933.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
