Assay Details
Gene Symbol: Zfp114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029933.2 | NP_001025104.2 | 2 - 3 | 160 | 151 | |
GenBank mRNA | BB633884.1 | - | 2 - 3 | 160 | 151 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001029933.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
