Assay Details
| Gene Symbol: Krt33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013570.1 | NP_038598.1 | 5 - 6 | 921 | 119 | |
| GenBank mRNA | AA762746.1 | - | 172 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P336049 | NM_013570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P336049 | AI325228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P336049 | X75650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P336049 | NM_013570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P336049 | AI325228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P336049 | X75650.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517750 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177333 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P336049 | NM_013570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P336049 | AI325228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P336049 | X75650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P336049 | NM_013570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P336049 | AI325228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P336049 | X75650.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07462601 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177333 | - | - |
| Whole Mouse Genome G4122A | A_51_P336049 | NM_013570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336049 | AI325228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336049 | X75650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336049 | NM_013570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336049 | AI325228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336049 | X75650.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09111442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09111432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065542 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390971 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390970 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427179_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390969 | - | - |
| Mouse Genome 430 2.0 Array | 1427179_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427179_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16671_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16671_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100034384.mm.1 | NM_013570.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034384.mm.1 | NM_013570.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034384.mm.1 | TC1100003749.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034385.mm.1 | NM_013570.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034385.mm.1 | NM_013570.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034385.mm.1 | TC1100003749.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034383.mm.1 | NM_013570.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034383.mm.1 | NM_013570.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034383.mm.1 | TC1100003749.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427179_at | - | - |
| Mouse Expression Set 430 | 1427179_PM_at | - | - |
| Murine Genome U74v2 Set | 97814_at | - | - |
| Murine Genome U74v2 Set | 162106_f_at | - | - |
| Murine Genome U74v2 Set | 162106_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97814_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427179_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427179_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427179_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427179_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 352517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2719704 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2719704 | - | - |

Krt33b
Genomic Map