Assay Details
| Gene Symbol: Rpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009081.2 | NP_033107.1 | 2 - 3 | 123 | 94 | |
| GenBank mRNA | AK012368.1 | - | 2 - 3 | 124 | 94 | |
| AK018714.1 | - | 2 - 3 | 124 | 94 | ||
| AK076133.1 | - | 2 - 3 | 124 | 94 | ||
| BC024395.1 | - | 2 - 3 | 123 | 4981342 | 94 | |
| BC086797.1 | - | 2 - 3 | 109 | 5713507 | 94 | |
| BC086932.1 | - | 2 - 3 | 125 | 6760499 | 94 | |
| BC092012.1 | - | 2 - 3 | 123 | 30608657 | 94 | |
| X74856.1 | - | 2 - 3 | 151 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl28
Genomic Map