Assay Details
| Gene Symbol: H2-D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC132088.1 | - | 1 - 2 | 163 | 40130365 | 135 |
| BC132655.1 | - | 1 - 2 | 163 | 40130932 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC132088.1, BC132655.1 transcripts.
Set Membership: |
|

H2-D1
Genomic Map