Assay Details
| Gene Symbol: Rpl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163485.1 | NP_001156957.1 | 1 - 2 | 93 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163485.1 transcripts.
Set Membership: |
|

Rpl30
Genomic Map