Assay Details
| Gene Symbol: Mgp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008597.3 | NP_032623.1 | 1 - 2 | 138 | 105 | |
| GenBank mRNA | AK018719.1 | - | 1 - 2 | 139 | 105 | |
| AK159591.1 | - | 1 - 2 | 142 | 105 | ||
| BC036991.1 | - | 1 - 2 | 134 | 4990627 | 105 | |
| BC069922.1 | - | 1 - 2 | 128 | 4984262 | 105 | |
| BC079478.1 | - | 1 - 2 | 129 | 4950210 | 105 | |
| BY702829.1 | - | 1 - 2 | 139 | 105 | ||
| D00613.1 | - | 1 - 2 | 81 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018719.1, AK159591.1, BC036991.1, BC069922.1, BC079478.1, D00613.1, NM_008597.3 transcripts.
Set Membership: |
|

Mgp
Genomic Map