Assay Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011186.1 | NP_035316.1 | 2 - 3 | 507 | 153 | |
| GenBank mRNA | AB003304.1 | - | 2 - 3 | 510 | 153 | |
| AF060091.1 | - | 2 - 3 | 507 | 153 | ||
| AK010441.1 | - | 2 - 3 | 527 | 153 | ||
| AK133839.1 | - | 2 - 3 | 528 | 153 | ||
| BC012246.1 | - | 2 - 3 | 515 | 4167355 | 153 | |
| BC106144.1 | - | 2 - 3 | 510 | 4953887 | 153 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
response to oxidative stress
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
proteolysis
response to oxidative stress
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613302 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613306 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613303 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613305 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613300 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613301 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115068 | BC106144.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115068 | BC012246.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115068 | AB003304.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115068 | AF060091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115068 | AK010441.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144850 | BC106144.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144850 | BC012246.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144850 | AB003304.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144850 | AF060091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144850 | AK010441.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83187 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149377 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144850 | BC106144.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144850 | BC012246.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144850 | AB003304.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144850 | AF060091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144850 | AK010441.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115068 | BC106144.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115068 | BC012246.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115068 | AB003304.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115068 | AF060091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115068 | AK010441.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269728 | BC106144.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269728 | BC012246.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269728 | AB003304.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269728 | AF060091.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269728 | AK010441.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149377 | - | - |
| Whole Mouse Genome G4122A | A_51_P269728 | BC106144.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269728 | BC012246.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269728 | AB003304.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269728 | AF060091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269728 | AK010441.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125168 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149377 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11383824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11535907 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11535901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11155726 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11383827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11535910 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11603518 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11535905 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11383830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613306 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613303 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613302 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613305 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613301 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11228279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11383834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11732662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11383832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11383827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11155726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11335653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11335655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11211800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613320 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419811 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415676_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419810 | - | - |
| Mouse Genome 430 2.0 Array | 1415676_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415676_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400015507.mm.1 | NM_011186.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015507.mm.1 | NM_011186.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015507.mm.1 | TC1400002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015508.mm.1 | NM_011186.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015508.mm.1 | NM_011186.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015508.mm.1 | TC1400002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015505.mm.1 | NM_011186.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015505.mm.1 | NM_011186.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015505.mm.1 | NR_030457.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015505.mm.1 | NR_030457.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015505.mm.1 | TC1400002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015504.mm.1 | NM_011186.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015504.mm.1 | NM_011186.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015504.mm.1 | TC1400002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015503.mm.1 | NM_011186.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015503.mm.1 | NM_011186.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015503.mm.1 | TC1400002061.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415676_a_at | - | - |
| Mouse Expression Set 430 | 1415676_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415676_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415676_a_at | - | - |
| Murine Genome U74v2 Set | 101558_s_at | - | - |
| Murine Genome U74v2 Set | 160559_PM_at | - | - |
| Murine Genome U74v2 Set | 160559_at | - | - |
| Murine Genome U74v2 Set | 101741_PM_at | - | - |
| Murine Genome U74v2 Set | 101558_PM_s_at | - | - |
| Murine Genome U74v2 Set | 101741_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415676_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415676_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415676_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415676_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415676_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415676_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 891684 | - | - |
| ABI Mouse Genome Survey Microarray | 894189 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6290242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290242 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290242 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613469 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613469 | - | - |
| mouse-6 v1.1 (Illumina) | scl019173.2_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290242 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019173.2_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290242 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613469 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613469 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019173.2_9-S | - | - |

Psmb5
Genomic Map