Assay Details
| Gene Symbol: Acot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019736.3 | NP_062710.2 | 7 - 8 | 715 | 96 | |
| GenBank mRNA | AJ238893.1 | - | 7 - 8 | 670 | 96 | |
| AK002892.1 | - | 7 - 8 | 679 | 96 | ||
| AK009717.1 | - | 7 - 8 | 678 | 96 | ||
| AK076513.1 | - | 8 - 9 | 769 | 96 | ||
| AK088082.1 | - | 7 - 8 | 677 | 96 | ||
| AK168713.1 | - | 7 - 8 | 717 | 96 | ||
| AK170876.1 | - | 7 - 8 | 678 | 96 | ||
| BC021763.1 | - | 7 - 8 | 705 | 4458575 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot9
Genomic Map