Assay Details
| Gene Symbol: Hax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011826.4 | NP_035956.1 | 2 - 3 | 456 | 122 | |
| GenBank mRNA | AF023482.1 | - | 2 - 3 | 428 | 122 | |
| AK002256.1 | - | 2 - 3 | 407 | 122 | ||
| AK010633.1 | - | 2 - 3 | 440 | 122 | ||
| AK088746.1 | - | 2 - 3 | 438 | 122 | ||
| BC006688.1 | - | 2 - 3 | 385 | 3582652 | 122 | |
| BC098225.1 | - | 2 - 3 | 410 | 1494110 | 122 | |
| BC147481.1 | - | 2 - 3 | 387 | 9056226 | 122 | |
More Information
Set Membership: |
|

Hax1
Genomic Map