Assay Details
| Gene Symbol: Foxi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101464.1 | NP_001094934.1 | 1 - 2 | 592 | 172 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxi3| Gene Symbol: Foxi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101464.1 | NP_001094934.1 | 1 - 2 | 592 | 172 | |
Set Membership: |
|