Assay Details
| Gene Symbol: Ddx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025860.3 | NP_080136.2 | 6 - 7 | 1141 | 89 | |
| GenBank mRNA | AK019845.1 | - | 6 - 7 | 1065 | 89 | |
| AK075864.1 | - | 6 - 7 | 1078 | 89 | ||
| BC017160.1 | - | 6 - 7 | 1061 | 89 | ||
| BC028246.1 | - | 6 - 7 | 1112 | 2582151 | 89 | |
| BC103776.1 | - | 6 - 7 | 1078 | 6492142 | 89 | |
| BC141230.1 | - | 6 - 7 | 1080 | 9055842 | 89 | |
| BC141231.1 | - | 6 - 7 | 1080 | 9055843 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ddx18
Genomic Map