| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10437453 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10437452 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1424871_PM_s_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10437451 |
- |
- |
| Mouse Genome 430 2.0 Array |
1424871_PM_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1424871_s_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
207740_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
207740_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1600009115.mm.1 |
NM_145359.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600009115.mm.1 |
NM_145359.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600009115.mm.1 |
TC1600001143.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600009117.mm.1 |
NM_145359.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600009117.mm.1 |
NM_145359.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600009117.mm.1 |
TC1600001143.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600009116.mm.1 |
NM_145359.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600009116.mm.1 |
NM_145359.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600009116.mm.1 |
TC1600001143.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1424871_PM_s_at |
- |
- |
| Mouse Expression Set 430 |
1424871_s_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1424871_PM_s_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1424871_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1424871_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1424871_PM_s_at |
- |
- |