Assay Details
| Gene Symbol: Rpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC081450.1 | - | 1 - 2 | 123 | 100 | |
| BC099380.1 | - | 1 - 2 | 121 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl41| Gene Symbol: Rpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC081450.1 | - | 1 - 2 | 123 | 100 | |
| BC099380.1 | - | 1 - 2 | 121 | 100 | ||
Set Membership: |
|