Assay Details
| Gene Symbol: Gm5916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167587.1 | NP_001161059.1 | 1 - 2 | 156 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5916| Gene Symbol: Gm5916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167587.1 | NP_001161059.1 | 1 - 2 | 156 | 116 | |
Set Membership: |
|