Assay Details
| Gene Symbol: Gpr137b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003568.1 | 7 | 2510 | 89 | ||
| GenBank mRNA | CN701965.1 | - | 1 | 144 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107207 | BC093499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107207 | AK009736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066301 | CN701965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066301 | DV659289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066301 | NR_003568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2187121 | DV659289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2187121 | AK171335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2187121 | AK170103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2187121 | NR_003568.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180800 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119488 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107207 | BC093499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107207 | AK009736.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10626482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10440424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10626489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10671463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10626466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28833531 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10407793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407794 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439256_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407792 | - | - |
| Mouse Genome 430 2.0 Array | 1429775_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450881_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1450881_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1450882_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1423164_at | - | - |
| Mouse Genome 430 2.0 Array | 1439256_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1450882_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429775_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423164_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439256_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 664862_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 664862_at | - | - |
| Mouse Expression Set 430 | 1439255_PM_s_at | - | - |
| Mouse Expression Set 430 | 1429775_a_at | - | - |
| Mouse Expression Set 430 | 1450881_PM_s_at | - | - |
| Mouse Expression Set 430 | 1439256_x_at | - | - |
| Mouse Expression Set 430 | 1423164_PM_at | - | - |
| Mouse Expression Set 430 | 1450882_s_at | - | - |
| Mouse Expression Set 430 | 1429775_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450881_s_at | - | - |
| Mouse Expression Set 430 | 1423164_at | - | - |
| Mouse Expression Set 430 | 1439255_s_at | - | - |
| Mouse Expression Set 430 | 1450882_PM_s_at | - | - |
| Mouse Expression Set 430 | 1439256_PM_x_at | - | - |
| Mouse Expression Set 430 | 1446806_at | - | - |
| Mouse Expression Set 430 | 1446806_PM_at | - | - |
| Murine Genome U74v2 Set | 103017_PM_at | - | - |
| Murine Genome U74v2 Set | 103017_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423164_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429775_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450881_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423164_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439256_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450882_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429775_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450881_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439256_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439255_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450882_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439255_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439256_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439256_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 766509 | - | - |
| ABI Mouse Genome Survey Microarray | 852384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2455850 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2455850 | - | - |

Gpr137b-ps
Genomic Map