| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10597620 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10597625 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10597622 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10597621 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10597619 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900027225.mm.1 |
NR_033303.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027225.mm.1 |
NR_033303.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027225.mm.1 |
TC0900003237.mm.1 |
MTA TC |