Assay Details
| Gene Symbol: Ugt2b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009467.3 | NP_033493.3 | 6 | 1543 | 153 | |
| GenBank mRNA | AI118071.1 | - | 1 | 153 | ||
| AK136480.1 | - | 6 | 1544 | 153 | ||
| X06358.1 | - | 6 | 1521 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975080 | X06358.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975080 | AK136480.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975080 | NM_009467.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9090 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975080 | X06358.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975080 | AK136480.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975080 | NM_009467.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P501007 | X06358.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P501007 | AK136480.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P501007 | NM_009467.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23866808 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05644325 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05644308 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01153657 | - | - |
| Whole Mouse Genome G4122A | A_52_P501007 | X06358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P501007 | AK136480.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P501007 | NM_009467.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113699 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05644308 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04884976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04884972 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04884979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04884972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04973245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04884973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04627841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04525256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23866771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531058 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419622_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531057 | - | - |
| Mouse Genome 430 2.0 Array | 1419622_at | - | - |
| Mouse Genome 430 2.0 Array | 1419622_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22238_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22238_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500023780.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023780.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023780.mm.1 | NM_009467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023774.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023774.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023774.mm.1 | NM_009467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023776.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023776.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023776.mm.1 | NM_009467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023775.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023775.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023775.mm.1 | NM_009467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023777.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023777.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023777.mm.1 | NM_009467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023778.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023778.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023778.mm.1 | NM_009467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023779.mm.1 | TC0500002693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023779.mm.1 | NM_009467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023779.mm.1 | NM_009467.3 | Refseq |
| Mouse Expression Set 430 | 1419622_at | - | - |
| Mouse Expression Set 430 | 1419622_PM_at | - | - |
| Murine Genome U74v2 Set | 96796_f_at | - | - |
| Murine Genome U74v2 Set | 96796_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419622_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419622_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419622_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419622_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 694979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2473505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2473505 | - | - |
| mouse-6 v1.1 (Illumina) | scl022238.1_263-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl022238.1_263-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2473505 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2473505 | - | - |

Ugt2b5
Genomic Map