Assay Details
| Gene Symbol: Igh-VJ558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF296437.1 | - | 1 | 139 | 138 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562158 | AF296435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562158 | AF296435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001281 | AB069917.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001281 | AB069913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001281 | AF296428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2140348 | BC103785.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121782 | AB069916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121782 | AF296424.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065506 | U58290.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103910 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150568 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118123 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149737 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158540 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172964 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105522 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104923 | AF296437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979364 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134277 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065506 | U58290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960148 | AK136471.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960148 | AI893585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960148 | AK002875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960148 | AK142609.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047974 | BC092049.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047974 | X73076.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2187030 | AF296436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513776 | AF296427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2140348 | BC103785.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC096617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC093501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC094065.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC096462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC058814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC096617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC093501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC094065.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC096462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509679 | BC058814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969901 | AF296432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270661 | - | - |
| Whole Mouse Genome G4122A | A_52_P139027 | U39781.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P686392 | AK007163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P686392 | AF296431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P686392 | BC053409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P686392 | AF045497.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P151887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09840016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09939537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10119877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10119882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10119871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09939647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09939630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10333618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09939538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28755686 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10402899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403051 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402900 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432520_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427852_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437346_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427839_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452574_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427850_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402864 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403048 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403015 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403079 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403054 | - | - |
| Mouse Genome 430 2.0 Array | 1452574_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427851_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427851_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427839_at | - | - |
| Mouse Genome 430 2.0 Array | 1427850_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427850_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427839_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432520_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452574_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427852_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427852_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1432520_at | - | - |
| Mouse Expression Set 430 | 1451947_PM_at | - | - |
| Mouse Expression Set 430 | 1427851_PM_x_at | - | - |
| Mouse Expression Set 430 | 1427850_PM_x_at | - | - |
| Mouse Expression Set 430 | 1427852_x_at | - | - |
| Mouse Expression Set 430 | 1427851_x_at | - | - |
| Mouse Expression Set 430 | 1429381_x_at | - | - |
| Mouse Expression Set 430 | 1427850_x_at | - | - |
| Mouse Expression Set 430 | 1421653_a_at | - | - |
| Mouse Expression Set 430 | 1425763_x_at | - | - |
| Mouse Expression Set 430 | 1427852_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452574_PM_x_at | - | - |
| Mouse Expression Set 430 | 1451949_at | - | - |
| Mouse Expression Set 430 | 1427839_at | - | - |
| Mouse Expression Set 430 | 1451949_PM_at | - | - |
| Mouse Expression Set 430 | 1421653_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451947_at | - | - |
| Mouse Expression Set 430 | 1426199_PM_x_at | - | - |
| Mouse Expression Set 430 | 1427839_PM_at | - | - |
| Mouse Expression Set 430 | 1426198_at | - | - |
| Mouse Expression Set 430 | 1451958_PM_at | - | - |
| Mouse Expression Set 430 | 1429381_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452574_x_at | - | - |
| Mouse Expression Set 430 | 1451958_at | - | - |
| Mouse Expression Set 430 | 1426198_PM_at | - | - |
| Mouse Expression Set 430 | 1425763_PM_x_at | - | - |
| Mouse Expression Set 430 | 1426199_x_at | - | - |
| Mouse Expression Set 430 | 1432520_at | - | - |
| Mouse Expression Set 430 | 1432520_PM_at | - | - |
| Murine Genome U74v2 Set | 97575_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97575_f_at | - | - |
| Murine Genome U74v2 Set | 100721_f_at | - | - |
| Murine Genome U74v2 Set | 102159_at | - | - |
| Murine Genome U74v2 Set | 101744_i_at | - | - |
| Murine Genome U74v2 Set | 102159_PM_at | - | - |
| Murine Genome U74v2 Set | 97568_at | - | - |
| Murine Genome U74v2 Set | 100375_PM_i_at | - | - |
| Murine Genome U74v2 Set | 99885_PM_at | - | - |
| Murine Genome U74v2 Set | 101321_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161486_f_at | - | - |
| Murine Genome U74v2 Set | 99421_PM_at | - | - |
| Murine Genome U74v2 Set | 100375_i_at | - | - |
| Murine Genome U74v2 Set | 93927_PM_f_at | - | - |
| Murine Genome U74v2 Set | 98776_PM_at | - | - |
| Murine Genome U74v2 Set | 99885_at | - | - |
| Murine Genome U74v2 Set | 100721_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101752_f_at | - | - |
| Murine Genome U74v2 Set | 100583_at | - | - |
| Murine Genome U74v2 Set | 161486_PM_f_at | - | - |
| Murine Genome U74v2 Set | 99829_at | - | - |
| Murine Genome U74v2 Set | 101744_PM_i_at | - | - |
| Murine Genome U74v2 Set | 101749_at | - | - |
| Murine Genome U74v2 Set | 101321_r_at | - | - |
| Murine Genome U74v2 Set | 101749_PM_at | - | - |
| Murine Genome U74v2 Set | 101320_f_at | - | - |
| Murine Genome U74v2 Set | 98776_at | - | - |
| Murine Genome U74v2 Set | 100362_f_at | - | - |
| Murine Genome U74v2 Set | 92470_f_at | - | - |
| Murine Genome U74v2 Set | 100682_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100583_PM_at | - | - |
| Murine Genome U74v2 Set | 101320_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97568_PM_at | - | - |
| Murine Genome U74v2 Set | 101752_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100362_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100682_f_at | - | - |
| Murine Genome U74v2 Set | 92470_PM_f_at | - | - |
| Murine Genome U74v2 Set | 99421_at | - | - |
| Murine Genome U74v2 Set | 99829_PM_at | - | - |
| Murine Genome U74v2 Set | 93927_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427850_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425763_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451958_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427839_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451949_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451949_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427850_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452574_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427851_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429381_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426198_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426198_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451958_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426199_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427852_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425763_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427839_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427851_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451947_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427852_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421653_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426199_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452574_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429381_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421653_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451947_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427839_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427839_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427852_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427852_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427850_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452574_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452574_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427850_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432520_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432520_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 60025 | - | - |

Igh-VJ558
Genomic Map